Learn More
Bachem Dermcidin-1L (human) Trifluoroacetate
SDP
Supplier:  Bachem 4061590.1000
Encompass_Preferred
Dermcidin-1L (human) Trifluoroacetate, H-Ser-Ser-Leu-Leu-Glu-Lys-Gly-Leu-Asp-Gly-Ala-Lys-Lys-Ala-Val-Gly-Gly-Leu-Gly-Lys-Leu-Gly-Lys-Asp-Ala-Val-Glu-Asp-Leu-Glu-Ser-Val-Gly-Lys-Gly-Ala-Val-His-Asp-Val-Lys-Asp-Val-Leu-Asp-Ser-Val-Leu-OH, 478898-18-9, C210H359N57O71, Mr 4818.50, 1mg. DCD-1L (human). The protein dermcidin expressed in the sweat glands is secreted into the sweat and transported to the epidermal surface. The concomitant processing of the protein yields a broad-spectrum antimicrobial 47 amino acid peptide, dermcidin-1 (DCD-1). Dermcidin and naturally occurring fragments as DCD-1L do not perturb membrane permeability and bind to distinct surface molecules. Niyonsaba et al. have reported that DCD-1L stimulates keratinocytes to generate cytokines and chemokines such as TNF-a, IL-8, CXCL10, and CCL20.
Catalog No. 50-194-6674