Learn More
Bachem Abaloparatide Acetate
SDP
Supplier:  Bachem 4105586.1000
Encompass_Preferred
Abaloparatide Acetate, H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Glu-Leu-Leu-Glu-Lys-Leu-Leu-Aib-Lys-Leu-His-Thr-Ala-NH2, 247062-33-5, C174H300N56O49, Mr 3960.64, 1mg. BIM-44058, BA 058, (Aib29)-Semparatide. The pTHrP-analog abaloparatide (BIM-44058) promoting bone growth has been developed for the treatment of osteoporosis.
Catalog No. 50-194-6976