Learn More
Enterprise Technology Solutions The Safety Zone Gloves,Latex Blend,Flock-lined,28 mil,Medium,1PR/BG,12 PR/DZ
SDP
Supplier:  Enterprise Technology Solutions SZNGRFOMD1SF
Encompass_Preferred
Neoprene Latex Blend Flock-lined Gloves may be universally known as "that dishwashing glove," but the uses extend beyond household cleaning. High-volume foodservice such as meat-processing plants benefit from their increased resistance to animal fats. Heavyweight construction with a 28 mil thick palm offers added chemical protection with greater chemical-resistance over standard latex. Gloves are 100 percent phthalate-free. Fish-scale grip on these 12" long gloves helps prevent slipping. Long, straight cuff protects your more than your hands.
Catalog No. 50-008-27813