Learn More
Bachem 2-Chlorotrityl chloride resin (100-200 mesh, 1.5-1.9 mmol/g)
SDP
Supplier:  Bachem 4040104.0025
Encompass_Preferred
2-Chlorotrityl chloride resin (100-200 mesh, 1.5-1.9 mmol/g), 934816-82-7, 25g. 2-ClTrt resin, 2-CTC resin.
Catalog No. 50-194-7370