Learn More
Perkin Elmer US LLC QUANTC Software for PerkinElmer Spectrum One FTIR
Supplier:  Perkin Elmer US LLC L1108758
Encompass_Preferred
The software also includes method validation, trend analysis, component correlation spectra and context-sensitive help. The software also includes method validation, trend analysis, component correlation spectra and context-sensitive help.This product can be used in conjunction with Spectrum Soft- ware, GRAMS Analyst, GRAMS Research, IRDM or as stand- alone package. NOTE: This software is supplied on compact disk only.
Catalog No. 50-201-6634