Learn More
Bachem Pasireotide Acetate
SDP
Supplier:  Bachem 4060221.0005
Encompass_Preferred
Pasireotide Acetate, Cyclo(-Hyp(2-aminoethyl-carbamoyl)-Phg-D-Trp-Lys-Tyr(Bzl)-Phe), 396091-73-9, C58H66N10O9, Mr 1047.22, 5mg. SOM230. The somatostatin analog pasireotide is a multi-receptor ligand with high binding affinity for somatostatin receptor subtypes sst(1,2,3) and sst(5). It suppresses ACTH secretion from pituitary adenomas. CAS-number (diacetate): 396091-77-3. Bachem provides this product solely for uses within the scope of any statute or law providing for an immunity, exemption, or exception to patent infringement (“Exempted Uses”), including but not limited to 35 U.S.C. § 271(e)(1) in the United States, the Bolar type exemption in Europe, and any corresponding exception to patent infringement in any other country.
Catalog No. 50-194-6726