Learn More
Bachem n-Octyltrioxyethylene
SDP
Supplier:  Bachem 4006522.0025
Encompass_Preferred
n-Octyltrioxyethylene, 19327-38-9, C14H30O4, Mr 262.39, 25g. C8E3, Triethyleneglycolmonooctylether. The tenside C8E3 solubilizes membrane proteins.
Catalog No. 50-194-7415