Learn More
Medchemexpress LLC Theaflavin-3'-gallate | 28543-07-9 | MFCD03427514 | 98.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN024410MG
Encompass_Preferred
Theaflavin-3'-gallate is a polyphenolic theaflavin monomer derived from black tea, supplied as a research reagent for biochemical and cellular studies. It has reported bioactivities including pro-oxidant effects and inhibition of xanthine oxidase, and is provided with safety documentation for laboratory handling.
  • High purity suitable for research use.
  • Available as small solid quantities and in DMSO solution for convenience.
  • Documented molecular data and CAS identifier for unambiguous identification.
  • Suitable for biochemical and cellular assays investigating oxidative stress and related pathways.
  • Provided with SDS/COA for safe handling and quality verification.
Catalog No. 50-002-95495