Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Theaflavin-3-gallate | 30462-34-1 | MFCD29918450 | 99.9% | 716.60 g/mol | C36H28O16 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN024510MG

Encompass_Preferred

Theaflavin-3-gallate is a polyphenolic theaflavin monomer derived from black tea, supplied as a research-grade reagent for studies of apoptosis, oxidative stress, and DNA damage signaling. It acts as a pro-oxidant in some cancer cell models and can induce p53-dependent cell-cycle arrest and DNA damage via the ATM/Chk/p53 pathway.

  • Purity 99.9%.
  • Molecular weight 716.60 g/mol.
  • Used to study apoptosis and p53-dependent mechanisms.
  • Induces oxidative stress and DNA damage in cell-based assays.
  • Available in small milligram sizes suitable for analytical work.
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-96447


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.