Learn More
TARGETMOL CHEMICALS INC THEAFLAVIN-3-GALLATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T30515MG
Encompass_Preferred
Also available in 10 mg 20 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Theaflavin-3-gallate (Theaflavine-3-Gallate) can play a role in decreased intestinal cholesterol absorption via inhibition of micelle formation. Theaflavin-3-gallate has anticancer and apoptotic effects in non-small cell lung carcinoma it acts as prooxidants and induce oxidative stress with carcinoma cells more sensitive than normal fibroblasts. purity: 99%
Catalog No. 50-257-7015