Learn More
Medchemexpress LLC HY-N0244 1mg Medchemexpress, Theaflavin-3'-gallate CAS:28543-07-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02441MG
Encompass_Preferred
Medchemexpress, HY-N0244 1mg Theaflavin-3'-gallate CAS:28543-07-9 Theaflavin-3'-gallate, a black tea theaflavin monomer, is regarded as the biologically important active component of black tea and provides health benefits. Theaflavin-3'-gallate acts as prooxidants and induces oxidative stress in the carcinoma cells. Theaflavin-3'-gallate reacts directly with reduced glutathione (GSH) in a time- and concentration-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2780