Learn More
TARGETMOL CHEMICALS INC Theaflavin 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T76025MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Theaflavin is a polyphenolic flavonoid that has been found in black tea and has diverse biological activities, including antioxidant, anticancer, anti-inflammatory, and antiviral properties Purity 99.28%
Catalog No. 50-287-7816