Learn More
Medchemexpress LLC HY-B0809 500mg Medchemexpress, Theophylline CAS:58-55-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0809500MG
Encompass_Preferred
Medchemexpress, HY-B0809 500mg Theophylline CAS:58-55-9 Theophylline is a nonselective phosphodiesterase (PDE) inhibitor, adenosine receptor blocker, and histone deacetylase (HDAC) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1127