Learn More
Wilmad Labglass THERM NON-MERC 10/30 -10/+250C
Supplier:  Wilmad Labglass LG10515152
Encompass
Thermometer, Celsius, Non-Mercury, with [ST] 10/30 Inner Joint. Temperature Range: -10degree to + 250degreeC; Length Below Joint: 280mm. Made with mineral spirits filling in white glass. With 1degreeC subdivisions.
Catalog No. 50-800-308