Learn More
GA International Thermal-transfer, xylene and solvent-resistant resin ribbons 3.15" x 984' / 80mm x 300m Black 1" / 25.4mm
Supplier:  GA International XAR80X300C11JZ4
Encompass_Preferred
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No. NC2478062