Learn More
Uline THERMA LABEL 2X11/2IN 8ROLL/CS
Supplier:  Uline S7411CS
Encompass_Preferred
Industrial Thermal Transfer Labels - 2 x 1 1⁄2", Ribbons Required3500 labels on rollDIMENSIONS:Roll Diameter (Actual): 7 1/8"Space Between Labels: 1/8"MATERIAL:Matte finish paper stockAdhesive: Permanent emulsion acrylicSHELF LIFE:1 yearTEMPERATURE:Application: 25° F and abovePerformance: -65° to 200° FCan be used in a freezer if applied and kept at room temperature for 24 hrs prior to freezing.
Catalog No. NC2917728