Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Thermal Transfer Printable Labels, B-424

Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of general purpose paper (B-424).

Supplier:  Brady™ THT254241

 View more versions of this product

Catalog No. 19-109-310


Add to Cart

Description

Description

  • For use with thermal transfer printers
  • Made of contrast and smear resistant paper
Specifications

Specifications

Recommended ribbon series: R4300; R4500; R6100
None
Matte
Paper (B-424)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
-40°C
1000 Label(s) / Roll
Rectangle
101.6 x 152.4 mm (W x H)
-40°C to 180°C
0.089 mm
6 in.
101.60 mm
6 in.
THT-25-424-1
Permanent Rubber
White
Roll
Single-side Printable
Thermal Transfer Printer Label
General and industrial identification
1
THT-25-424-1
1000
101.60 mm (W) x 152.40 mm (H)
-40°F to 365°F
0.0035 in.
101.60 mm (W) x 152.40 mm (H)
4 in.
101.60 mm
152.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.