Learn More
Cole-Parmer TSI AVM440-A CompuFlow Thermoanemometer, auto datalogging w/humidity
Supplier:  Cole-Parmer 1025036
Encompass
These thermoanemometers measure air velocity, air volume, and temperature. Display up to three measurements simultaneously on a large backlit LCD. Automatically calculate averages of all your parameters with the push of a button.
Catalog No. 50-633-418