Learn More
Inorganic Ventures THERMO TUNING SOLUTION 125mL
SDP
Supplier:  Inorganic Ventures THMTS1125ML
Encompass_Preferred
Inorganic Ventures' THM ICP-MS Tuning Solution is a 125mL certified reference material set in a nitric acid (tr. hydrochloric acid) matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-837