Learn More
Gentech Scientific Inc Refurbished Thermo Vantage
SDP
Supplier:  Gentech Scientific Inc GTS11333
Encompass_Preferred
• TSQ Vantage LC/MS/MS, • HESI II Source, • Two Mechanical Pumps, • Xcalibur Data System, • 30 Day Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions. Note: HPLC is not included, is an available option for additional purchase.
Catalog No. 50-189-7492
Only null left
Add to Cart
Request a Quote