Learn More
Chemglass Life Sciences Thermometer, Non-Mercury, 10/30 Joint, 76mm Immersion, -20°C to 150°C
SDP
Supplier:  Chemglass Life Sciences CG3506N03
Encompass_Preferred
Thermometer, Non-Mercury, 10/30 Joint, 76mm Immersion, -20¿C to 150¿C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. Thermometer has a white back, a 6 to 7mm outer diameter, and a standard taper 10/30 inner ground joint. All ranges have 1¿ subdivisions. Immersion is measured from the bottom of the joint to the bottom of the mercury bulb.
Catalog No. CG3506N03