Learn More
Vee Gee Thermometer, Dial, 50 to 550°F, 1.75 in Diam.
Supplier:  Vee Gee 82550/DEL
Encompass_Preferred
The dial thermometer indicates temperature through the metal stem with attached circular scale. It does don’t require batteries, it is rugged, and readings can be adjusted with the calibration screw on the back. • Larger dial size than common pocket thermometers for easier reading• Includes a clip for container mounting • Face Diameter: 1.75 in (44.5 mm), Stem Length: 8 in (203 mm), Stem Diameter: 0.138 in (3.5 mm)
Catalog No. NC0856578