Learn More
Thermco Products Inc NIST Traceable Digital Thermometer w/Probe. Use for Freezers, Refrigerators, Incubators, Water Baths, Block Heaters,and Ovens
Supplier: Thermco Products Inc ACC801DIG
Multi purpose digital thermometer, with minimum-maximum memory,display back light, celsius-fahrenheit exchange. Can be used to measure interal and external temperatures. Sensor cable is 3000mm long, display size 48mm x 21mm. Accuracy ±0.5 from -20° to +50°C, (±1° -4° to 122°F)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.