Learn More
Med Vet International McKesson Datalogging Refrigerator / Freezer Thermometer, Digital Display, MCK80022P, Each
Supplier:  Med Vet International 1074442EA
Encompass_Preferred
Datalogging Refrigerator / Freezer Thermometer with Alarm McKesson Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) 2 Glycol Bottle Probes / Internal Sensor Flip-out Stand Battery Operated Display shows current, min/max temperatures with a 10-second refresh rate, accuracy of ±0.5°C and a 0.1° display resolution Certified, serialized and traceable and has been tested against a standard calibrated at the National Institute of Standards and Technology (NIST) Waterproof, external bottle probe with 2000 mm cable monitors refrigerators or freezer temperatures from -22 - 158° F (-30 - 70° C) Includes internal sensor to measure room temperatures from 32 - 122° F (0 - 50° C) Logs Data once per minute to once per 720 min according to user preference Powered by (3) 1.5 V Type AA batteries (or equivalent) with a life of ~2000 hours in continuous operation when no alarm was triggered, and low-battery indications
Catalog No. 50-304-6371