Learn More
Brookfield Engineer Labs THERMOSEL/SSA SPINDLE NOSC4-31
Supplier:  Brookfield Engineer Labs SC431
Encompass
Spindle, For use with the Thermosel System, Selection is determined by the viscosity measurement range that is required
Catalog No. NC9683217