Learn More
Electron Microscopy Sciences Thermostatic Dryer
SDP
Supplier:  Electron Microscopy Sciences 74836
Encompass_Preferred
For RC and fiber base papers. It accommodates one 11x14", two 8x10" or eight 4x5" prints at once on each drying surface. Fast start; Quick drying; No over heating. Has maximum uniformity of surface heat. Made from a highly polished chrome Ferrotype metal. Drying surface: 12x17" (30.5x43.1cm)
Catalog No. 50-292-88