Learn More
Medchemexpress LLC Theviridoside | 23407-76-3 | 99.6% | 404.37 | C17H24O11 | 10 MG

Supplier: Medchemexpress LLC HYN115510MG
Theviridoside is a natural iridoid glucoside isolated from the leaves of Cerbera odollam with reported cytotoxic activity. It is supplied as a purified research reagent for biochemical and cellular studies.
- Purity 99.58%.
- Molecular formula C17H24O11, molecular weight 404.37 g·mol-1.
- Suitable for in vitro cytotoxicity and biochemical assays.
- Supplied as a 10 mg vial for research use.
- Store and handle according to laboratory safety practices.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.