Learn More
Cayman Chemical ThIacloprId-d4 5mg
SDP
Supplier:  Cayman Chemical 395735 MG
Encompass_Preferred
An internal standard for the quantification of thiacloprid by GC- or LC-MS
Catalog No. 50-291-8398