Learn More
Selleck Chemical LLC Thiamet G 25mg 1009816-48-1
SDP
Supplier:  Selleck Chemical LLC S721325MG
Encompass_Preferred
Thiamet G is a potent, selective O-GlcNAcase inhibitor with Kiof 21 nM, while exhibiting 37,000-fold selectivity over human lysosomal ¿hexosaminidase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4526