Learn More
Medchemexpress LLC HY-B0833 500mg Medchemexpress, Thiamethoxam CAS:153719-23-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0833500MG
Encompass_Preferred
Medchemexpress, HY-B0833 500mg Thiamethoxam CAS:153719-23-4 Thiamethoxam is a broad spectrum neonicotinoid insecticide[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1133