Learn More
Medchemexpress LLC Thiamethoxam (Standard) | 153719-23-4 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0833R50MG
Encompass_Preferred
Thiamethoxam (Standard) is the analytical standard of Thiamethoxam, a broad spectrum neonicotinoid insecticide. Intended for research and analytical applications, this product is a reference standard suitable for various assays.
  • Used as a reference standard for assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for techniques such as HPLC, GC, and MS.
Catalog No. 50-004-08059