Learn More
Medchemexpress LLC Thiamine-d3 (hydrochloride) | 2241872-28-4 | 340.29 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0680S11MG
Encompass_Preferred
Thiamine-d3 hydrochloride is the deuterium labeled Thiamine hydrochloride, an essential micronutrient that functions as a cofactor for many central metabolic enzymes. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16707