Learn More
Selleck Chemical LLC Thiamine HCl (Vitamin B1) S3211-200mg
SDP
Supplier:  Selleck Chemical LLC S3211200MG
Encompass_Preferred
Thiamine or vitamin B1 is a water-soluble vitamin of the B complex its phosphate derivatives are involved in many cellular processes
Catalog No. 50-311-0502