Learn More
Apexbio Technology LLC Thiamine HCl (Vitamin B1) 67-03-8 10mM (in 1mL H20)
SDP
Supplier:  Apexbio Technology LLC B18525.1
Encompass_Preferred
Thiamine HCl (Vitamin B1) (67-03-8) is a water-soluble micronutrient and biological precursor involved in coenzyme-dependent metabolic pathways It undergoes phosphorylation in cells to form thiamine pyrophosphate (TPP) which participates in carbohydrate metabolism through enzymatic reactions involving pyruvate dehydrogenase alpha-ketoglutarate dehydrogenase and branched-chain alpha-keto acid dehydrogenase complexes Thiamine HCl exerts its biological activity primarily through serving as a precursor to the coenzyme TPP enabling regulation of carbohydrate metabolic pathways In vitro assays frequently apply thiamine HCl to investigate enzyme activity regulation and coenzyme-dependent processes however clearly reported IC50 values are not specified due to its principal function as a coenzyme precursor Based on these attributes thiamine HCl holds research potential in studies of cellular energetics neuronal function and metabolic processes
Catalog No. 50-000-88561