Learn More
Medchemexpress LLC Thiamine hydrochloride | 67-03-8 | MFCD00012780 | 337.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0680R50MG
Encompass_Preferred
Thiamine hydrochloride (Standard) is an analytical standard and essential micronutrient, functioning as a cofactor for various central metabolic enzymes. This product is designed for research and analytical applications.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Essential micronutrient and cofactor for metabolic enzymes.
Catalog No. 50-003-89109