Learn More
Medchemexpress LLC Thiamine hydrochloride | 67-03-8 | MFCD00012780 | 99.8% | 337.27 | C12H18Cl2N4OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0680R10MG
Encompass_Preferred
Thiamine hydrochloride (vitamin B1) analytical standard provided as the hydrochloride salt for use as a reference material in research and analytical applications. It is a water-soluble vitamin used in studies of metabolic enzymes and related biochemical pathways.
  • Intended for use as a reference standard in analytical and research applications.
  • High purity suitable for analytical work (99.8%).
  • Molecular weight 337.27 and formula C12H18Cl2N4OS.
  • Supplied in a small reference quantity (10 mg) for assay calibration and method validation.
  • Water-soluble vitamin B1 useful in metabolic, nutritional, and biochemical assays.
Catalog No. 50-002-96953