Learn More
Medchemexpress LLC Thiamine hydrochloride (Standard) | 67-03-8 | 337.27 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0680R100MG
Encompass_Preferred
Thiamine hydrochloride (Standard) is the analytical standard of Thiamine (hydrochloride), intended for research and analytical applications. Thiamine hydrochloride is an essential micronutrient needed as a cofactor for many central metabolic enzymes. It is commonly used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Essential micronutrient and cofactor for metabolic enzymes.
  • Used as a reference for assay.
  • Suitable for HPLC, GC, and MS experiments.
Catalog No. 50-003-88962