Learn More
Alkali Scientific Thiamine Hydrochloride [Vitamin B1], 250 G
SDP
Supplier:  Alkali Scientific NEH004CP
Encompass_Preferred
Thiamine Hydrochloride (Vitamin B1) is an essential water-soluble vitamin used in biochemical research and clinical applications. This 250g package is ideal for studies involving cellular metabolism, particularly in the conversion of carbohydrates into energy. Thiamine plays a critical role in maintaining proper nerve function and cardiovascular health. It is often used in research on metabolic disorders, neurological diseases, and vitamin B1 deficiency conditions, such as beriberi. This reagent is also utilized in the formulation of therapeutic products for treating thiamine deficiency and related diseases. Thiamine Hydrochloride ensures precise and consistent results in scientific experiments and clinical assays.
Catalog No. 50-006-61272