Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamine monochloride | 59-43-8 | 99.96% | C12H17ClN4OS | 1 G
SDP

Supplier:  Medchemexpress LLC HYA01001G

Encompass_Preferred

Thiamine monochloride (Vitamin B1) is an essential vitamin playing a critical role in cellular energy production and neuronal activities. This solid substance is primarily used as a laboratory chemical and for manufacturing other substances. It is stable under recommended storage conditions and has a molecular weight of 300.81.

  • Essential vitamin for cellular energy production
  • Enhances normal neuronal activities
  • Primarily used as a laboratory chemical
  • Stable under recommended storage conditions
  • White to off-white solid appearance
  • Purity of 99.96%
  • Molecular weight of 300.81

Catalog No. 50-004-32784


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.