Learn More
Medchemexpress LLC Thiamine monochloride (Vitamin B1) | 59-43-8 | 99.96% | C12H17ClN4OS | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA010010MM/1ML
Encompass_Preferred
Thiamine monochloride (Vitamin B1) is an essential vitamin that plays an important role in cellular production of energy from ingested food and enhances normal neuronal activities. It is for research use only.
  • Essential vitamin
  • Plays a role in cellular energy production
  • Enhances normal neuronal activities
  • High purity: 99.96%
  • Available in both solid and solution forms
Catalog No. 50-004-32119