Learn More
Medchemexpress LLC Thiamine monophosphate chloride (dihydrate) | 273724-21-3 | 99.2% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1287421G
Encompass_Preferred
Thiamine monophosphate chloride dihydrate is a vitamin B1 metabolite and thiamine precursor. This compound is absorbable in vivo and can be converted into thiamine within the body. It is mainly used in research as a vitamin B1 source and can also function as a food additive.
  • Vitamin B1 metabolite
  • Thiamine precursor
  • Absorbable in vivo
  • Converted into thiamine in the body
  • Can be used as a food additive
  • Source of vitamin B1 for research and food supplements
Catalog No. 50-003-39995