Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamine nitrate | 532-43-4 | 99.41% | 1 G
SDP

Supplier:  Medchemexpress LLC HYB22231G

Encompass_Preferred

Thiamine nitrate is an essential vitamin that can enhance normal neuronal activities. It is for research use only and not sold to patients.

  • High purity of 99.41%.
  • Appears as a solid, white to off-white in color.
  • Store at 4°C, sealed, away from moisture and light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO at 20 mg/mL.
  • Used in studies concerning neuronal activities and thiamine-restricted diets.

Catalog No. 50-004-10216


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.