Learn More
Medchemexpress LLC Thiamine nitrate | 532-43-4 | 99.41% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB222310G
Encompass_Preferred
Thiamine nitrate, also known as Vitamin B1 nitrate, is a vital vitamin that plays a crucial role in enhancing normal neuronal activity. This compound is extensively used in research to study thiamine deficiency and its neurological impacts.
  • Enhances normal neuronal activity.
  • Essential vitamin.
  • Used in research for studying thiamine deficiency.
  • Impacts observed in in vivo studies include effects on paralysis and weight loss.
  • Supports sustained survival in models fed thiamine-restricted diets.
Catalog No. 50-004-10953