Learn More
Cayman Chemical Thiamine Pyrophosphate chlor
SDP
Supplier:  Cayman Chemical 20254500 MG
Encompass_Preferred
A metabolite of vitamin B1; a cofactor synthesized in the cytosol; required for the activity of cytosolic transketolase and mitochondrial pyruvate, oxoglutarate, and branched-chain keto acid dehydrogenases; prevents hyperglycemia-induced retinopathy and desflurane-induced hepatotoxicity in rats
Catalog No. 50-298-4386