Learn More
Medchemexpress LLC Thiamine pyrophosphate (cocarboxylase) | 154-87-0 | 99.9% | 460.77 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY113076500MG
Encompass_Preferred
Thiamine pyrophosphate is the coenzyme form of Vitamin B1, and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. It is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Is the coenzyme form of Vitamin B1.
  • Required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex.
  • Necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Inhibits ischemia-reperfusion induced oxidative damage in rat ovarian tissue.
  • Prevents Cisplatin-induced cardiotoxicity, and associated oxidative stress and heart injury in rats.
Catalog No. 50-004-37172