Learn More
Medchemexpress LLC Thiamine pyrophosphate (Standard) | 154-87-0 | 98.4% | 460.77 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY113076R
Encompass_Preferred
Thiamine pyrophosphate (Standard) is the analytical standard of Thiamine pyrophosphate, intended for research and analytical applications. It is the coenzyme form of Vitamin B1 and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. It is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Coenzyme form of vitamin B1
  • Required intermediate in metabolic complexes
  • Necessary for oxidative phosphorylation and pentose phosphate pathway
  • Cofactor for α-ketoacid dehydrogenases
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-39552