Learn More
Apexbio Technology LLC Thiamphenicol 15318-45-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18535.1
Encompass_Preferred
Thiamphenicol (CAS 15318-45-3) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to interfere with bacterial protein synthesis thereby regulating translational processes Thiamphenicol exerts its biological activity primarily through inhibition of peptide elongation by binding to the 50S ribosomal subunit In experimental studies Thiamphenicol demonstrates bacteriostatic activity with reported IC50 values for bacterial strains typically ranging from 0 5 8 g/mL depending on organism sensitivity growth conditions and assay methodologies Based on these pharmacological properties Thiamphenicol holds research potential in studying bacterial resistance mechanisms screening antibiotic candidates and investigating translation regulation in microbial systems
Catalog No. 50-000-90314