Learn More
Cayman Chemical ThIamphenIcol 50g
SDP
Supplier:  Cayman Chemical 2135750 G
Encompass_Preferred
An antibiotic derived from chloramphenicol that displays a similar spectrum of activity with enhanced potency
Catalog No. 50-298-0891