Learn More
Medchemexpress LLC Thiamphenicol-d3 | 2211914-19-9 | 99.5% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0479S1MG
Encompass_Preferred
Thiamphenicol-d3 is a deuterium labeled form of Thiamphenicol, a broad-spectrum antimicrobial antibiotic. As a methyl-sulfonyl derivative of Chloramphenicol, it functions by binding to the 50S ribosomal subunit, which inhibits protein synthesis. This mechanism provides a bacteriostatic effect against a range of bacteria, including Gram-negative, Gram-positive aerobic, and anaerobic types.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06720