Learn More
Medchemexpress LLC Thiamutilin (fumarate) | 55297-96-6 | 99.27% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB2060A
Encompass_Preferred
Tiamulin fumarate is a diterpenic antibiotic widely used in pigs and poultry to control infectious diseases. It is effective in the study of airsacculitis, primarily caused by Mycoplasma spp. It is a semisynthetic derivative of the diterpene antibiotic Pleuromutilin and binds to rRNA to inhibit protein production.
  • Used in pigs and poultry for infectious disease control
  • Effective in studying airsacculitis caused by Mycoplasma spp.
  • Highly active in vitro against Mycoplasma strains, Spirochaetes, and gram-positive bacteria
  • Less active against gram-negative bacteria
  • Binds to rRNA, inhibiting protein production
  • Highly effective in treating avian intestinal spirochaetosis in breeder and layer hens
Catalog No. 50-004-14436